missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3,5-dimethylbenzoic acid, 98+%

Catalog No. p-7046485
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1453603 1 g
AAL1453606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1453603 Supplier Thermo Scientific Chemicals Supplier No. L1453603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97888-80-7 | C16H16O3 | 256.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97888-80-7
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00272572
InChI Key JABUPJCJZZNUFK-UHFFFAOYSA-N
Synonym 4-benzyloxy-3,5-dimethylbenzoic acid, 4-benzyloxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl-4-phenylmethoxy benzoic acid, 4-benzyloxy-3,5-dimethyl-benzoate
PubChem CID 2733993
IUPAC Name 3,5-dimethyl-4-phenylmethoxybenzoic acid
SMILES CC1=CC(=CC(=C1OCC2=CC=CC=C2)C)C(=O)O

Specifications

Melting Point 157°C to 160°C
Quantity 1 g
Beilstein 5045861
Formula Weight 256.3
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxy-3,5-dimethylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only