missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxy-2-bromopropiophenone, 97%, Thermo Scientific Chemicals

Catalog No. p-7057170
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6657814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6657814 Supplier Thermo Scientific Chemicals Supplier No. H6657814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4'-Benzyloxy-2-bromopropiophenone is an intermediate in the preparation of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Benzyloxy-2-bromopropiophenone is an intermediate in the preparation of Bazedoxifene acetate.

Solubility
Soluble in water (2.342g/L at 25°C ).

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 35081-45-9
Molecular Formula C16H15BrO2
Molecular Weight (g/mol) 319.198
MDL Number MFCD00017862
InChI Key LSQLSCUJBXFBRB-UHFFFAOYSA-N
Synonym 4'-benzyloxy-2-bromopropiophenone, 2-bromo-4'-benzyloxypropiophenone, 1-4-benzyloxy phenyl-2-bromopropan-1-one, 1-propanone, 2-bromo-1-4-phenylmethoxy phenyl, 4-benzyloxy-2-bromopropiophenone, 1-4-benzyloxyphenyl-2-bromo-propan-1-one, 1-4-benzyloxy-phenyl-2-bromo-propan-1-one, 1-4-benzyloxy phenyl-2-bromo-1-propanone
PubChem CID 100040
IUPAC Name 2-bromo-1-(4-phenylmethoxyphenyl)propan-1-one
SMILES CC(C(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2)Br

Specifications

Melting Point 71°C to 76°C
Quantity 25 g
Beilstein 1882887
Solubility Information Soluble in water (2.342g/L at 25°C ).
Formula Weight 319.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4'-Benzyloxy-2-bromopropiophenone
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 252-351-6
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only