CAS RN 35081-45-9
IUPAC Name:
1-[4-(benzyloxy)phenyl]-2-bromopropan-1-one
Synonyms:
1-[4-(benzyloxy)phenyl]-2-bromopropan-1-one
Molecular Weight (g/mol):
319.2
Molecular Formula:
C16H15BrO2
InChi Key:
LSQLSCUJBXFBRB-UHFFFAOYNA-N
SMILES:
CC(Br)C(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4'-Benzyloxy-2-bromopropiophenone, 97%, Thermo Scientific Chemicals
CAS: 35081-45-9 Molecular Formula: C16H15BrO2 Molecular Weight (g/mol): 319.198 MDL Number: MFCD00017862 InChI Key: LSQLSCUJBXFBRB-UHFFFAOYSA-N Synonym: 4'-benzyloxy-2-bromopropiophenone,2-bromo-4'-benzyloxypropiophenone,1-4-benzyloxy phenyl-2-bromopropan-1-one,1-propanone, 2-bromo-1-4-phenylmethoxy phenyl,4-benzyloxy-2-bromopropiophenone,1-4-benzyloxyphenyl-2-bromo-propan-1-one,1-4-benzyloxy-phenyl-2-bromo-propan-1-one,1-4-benzyloxy phenyl-2-bromo-1-propanone PubChem CID: 100040 IUPAC Name: 2-bromo-1-(4-phenylmethoxyphenyl)propan-1-one SMILES: CC(C(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2)Br