missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylbiphenyl 99.0+%, TCI America™

Catalog No. p-7119090
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15195G 5 g
B151925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15195G Supplier TCI America Supplier No. B15195G

Chemical Identifiers

CAS 613-42-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00041831
InChI Key AGPLQTQFIZBOLI-UHFFFAOYSA-N
Synonym 4-benzylbiphenyl, 4-phenylmethyl-1,1'-biphenyl, 1,1'-biphenyl, 4-phenylmethyl, benzylbiphenyl, p-benzylbiphenyl, 4-phenyldiphenylmethane, 1-phenyl-4-benzylbenzene, 1, 4-phenylmethyl, acmc-1bh4u, 1-benzyl-4-phenyl-benzene
PubChem CID 69174
IUPAC Name 4-benzyl-1,1'-biphenyl
SMILES C(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 86°C
Color White
Boiling Point 286°C
Quantity 5 g
Formula Weight 244.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzylbiphenyl
EINECSNumber (4)-1501
TSCA No