Learn More
4-Benzyl-3-thiosemicarbazide, 98+%
CAS: 13431-41-9 | C8H11N3S | 181.257 g/mol
$101.15 - $284.22
Chemical Identifiers
| CAS | 13431-41-9 |
|---|---|
| Molecular Formula | C8H11N3S |
| Molecular Weight (g/mol) | 181.257 |
| MDL Number | MFCD00025145 |
| InChI Key | ZTRUHAVBRPABTK-UHFFFAOYSA-N |
| Synonym | 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea |
| PubChem CID | 737135 |
| IUPAC Name | 1-amino-3-benzylthiourea |
| SMILES | C1=CC=C(C=C1)CNC(=S)NN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1092003
|
Thermo Scientific Chemicals
L1092003 |
1 g |
N/A
|
|
|||||
|
AAL1092006
|
Thermo Scientific Chemicals
L1092006 |
5 g |
N/A
|
|
|||||
Description
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
| 13431-41-9 | |
| 181.257 | |
| ZTRUHAVBRPABTK-UHFFFAOYSA-N | |
| 737135 | |
| C1=CC=C(C=C1)CNC(=S)NN |
| C8H11N3S | |
| MFCD00025145 | |
| 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea | |
| 1-amino-3-benzylthiourea |
Specifications
| 13431-41-9 | |
| C8H11N3S | |
| 1 g | |
| 2096431 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)CNC(=S)NN | |
| 181.257 | |
| 181.26 | |
| 4-Benzyl-3-thiosemicarbazide |
| 128°C to 131°C | |
| MFCD00025145 | |
| UN2811 | |
| 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea | |
| ZTRUHAVBRPABTK-UHFFFAOYSA-N | |
| 1-amino-3-benzylthiourea | |
| 737135 | |
| ≥98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only