missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-3-thiosemicarbazide, 98+%

Catalog No. AAL1092003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1092003 1 g
AAL1092006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1092003 Supplier Thermo Scientific Chemicals Supplier No. L1092003

CAS: 13431-41-9 | C8H11N3S | 181.257 g/mol

4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 13431-41-9
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.257
MDL Number MFCD00025145
InChI Key ZTRUHAVBRPABTK-UHFFFAOYSA-N
Synonym 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea
PubChem CID 737135
IUPAC Name 1-amino-3-benzylthiourea
SMILES C1=CC=C(C=C1)CNC(=S)NN

Specifications

Melting Point 128°C to 131°C
Quantity 1 g
UN Number UN2811
Beilstein 2096431
Solubility Information Insoluble in water.
Formula Weight 181.26
Percent Purity ≥98%
Chemical Name or Material 4-Benzyl-3-thiosemicarbazide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only