missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-morpholinecarboxylic acid hydrochloride, 97%, Thermo Scientific™

Catalog No. p-2701489
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12401CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12401CB Supplier Thermo Scientific Chemicals Supplier No. CC12401CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135072-15-0
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.714
MDL Number MFCD02682031
InChI Key CEDXMALCJZSQHA-UHFFFAOYSA-N
Synonym 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
PubChem CID 2776352
IUPAC Name 4-benzylmorpholine-2-carboxylic acid;hydrochloride
SMILES C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl

Specifications

CAS Min % 97
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 257.72
Percent Purity 97%
Chemical Name or Material 4-Benzyl-2-morpholinecarboxylic acid hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.