missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Benzyl-1,4-oxazinan-2-yl)methanol, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12409DE
Specifications
| (4-Benzyl-1, 4-oxazinan-2-yl)methanol | |
| 94.5 | |
| 95% | |
| MFCD02682009,MFCD10698663,MFCD11848685 | |
| 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw | |
| OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1 | |
| 208.28 | |
| 207.27 |
| 40987-24-4 | |
| 100.0 | |
| C12H18NO2 | |
| 5g | |
| WQNIKIMRIXHNFF-GFCCVEGCSA-O | |
| (2R)-4-benzyl-2-(hydroxymethyl)morpholin-4-ium | |
| 2776358 | |
| 95% |
Chemical Identifiers
| 40987-24-4 | |
| 208.28 | |
| WQNIKIMRIXHNFF-GFCCVEGCSA-O | |
| 2776358 | |
| OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1 |
| C12H18NO2 | |
| MFCD02682009,MFCD10698663,MFCD11848685 | |
| 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw | |
| (2R)-4-benzyl-2-(hydroxymethyl)morpholin-4-ium |