missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Benzyl-1,4-oxazinan-2-yl)methanol, Thermo Scientific™
Chemical Identifiers
| CAS | 40987-24-4 |
|---|---|
| Molecular Formula | C12H18NO2 |
| Molecular Weight (g/mol) | 208.28 |
| MDL Number | MFCD02682009,MFCD10698663,MFCD11848685 |
| InChI Key | WQNIKIMRIXHNFF-GFCCVEGCSA-O |
| Synonym | 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw |
| PubChem CID | 2776358 |
| IUPAC Name | (4-benzylmorpholin-2-yl)methanol |
| SMILES | OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
CC12409CB
|
Thermo Scientific Chemicals
CC12409CB |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC12409DA
|
Thermo Scientific Chemicals
CC12409DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC12409DE
|
Thermo Scientific Chemicals
CC12409DE |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 40987-24-4 | |
| 208.28 | |
| WQNIKIMRIXHNFF-GFCCVEGCSA-O | |
| 2776358 | |
| OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1 |
| C12H18NO2 | |
| MFCD02682009,MFCD10698663,MFCD11848685 | |
| 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw | |
| (4-benzylmorpholin-2-yl)methanol |
Specifications
| 40987-24-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD02682009,MFCD10698663,MFCD11848685 | |
| 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw | |
| OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1 | |
| 208.28 | |
| 207.27 | |
| (4-Benzyl-1, 4-oxazinan-2-yl)methanol |
| 94.5 | |
| 95% | |
| C12H18NO2 | |
| 250mg | |
| WQNIKIMRIXHNFF-GFCCVEGCSA-O | |
| (4-benzylmorpholin-2-yl)methanol | |
| 2776358 | |
| 95% |