missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbiphenyl, 99%

Catalog No. AAA1557314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1557314 25 g
AAA1557322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1557314 Supplier Thermo Scientific Chemicals Supplier No. A1557314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2128-93-0 | C19H14O | 258.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2128-93-0
Molecular Formula C19H14O
Molecular Weight (g/mol) 258.32
MDL Number MFCD00003079
InChI Key LYXOWKPVTCPORE-UHFFFAOYSA-N
Synonym 4-benzoylbiphenyl, 4-phenylbenzophenone, p-phenylbenzophenone, benzophenone, 4-phenyl, p-benzoylbiphenyl, 4-diphenyl phenyl ketone, p-biphenylyl phenyl ketone, trigonal 12, 4-biphenylyl phenyl ketone, methanone, 1,1'-biphenyl-4-ylphenyl
PubChem CID 75040
IUPAC Name phenyl-(4-phenylphenyl)methanone
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 98°C to 103°C
Quantity 25 g
Beilstein 1876092
Formula Weight 258.32
Percent Purity 99%
Chemical Name or Material 4-Benzoylbiphenyl
EINECSNumber 218-345-2
RTECSNumber PC4936800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only