missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoyl-L-phenylalanine, 95%

CAS: 104504-45-2 | C16H15NO3 | 269.30 g/mol

Supplier:  Thermo Scientific Chemicals H5208306

 View more versions of this product

Catalog No. AAH5208306


Only null left
Explore available promotions

Description

Description

4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Benzoyl-L-phenylalanine
C16H15NO3
s-2-amino-3-4-benzoylphenyl propanoic acid, l-4-benzoylphenylalanine, para-benzoyl-phenylalanine, h-bpa-oh h-phe 4-bz-oh, 4-benzoyl-l-phenylalanine, 4-benzoylphenylalanine, l-p-benzoylphenylalanine, phenylalanine, 4-benzoyl, h-bpa-oh, h-p-bz-phe-oh
N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O
5 g
7020128
95%
104504-45-2
MFCD00238102
TVIDEEHSOPHZBR-AWEZNQCLSA-N
(2S)-2-amino-3-(4-benzoylphenyl)propanoic acid
269.30
269.3

Chemical Identifiers

104504-45-2
269.30
TVIDEEHSOPHZBR-AWEZNQCLSA-N
7020128
N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O
C16H15NO3
MFCD00238102
s-2-amino-3-4-benzoylphenyl propanoic acid, l-4-benzoylphenylalanine, para-benzoyl-phenylalanine, h-bpa-oh h-phe 4-bz-oh, 4-benzoyl-l-phenylalanine, 4-benzoylphenylalanine, l-p-benzoylphenylalanine, phenylalanine, 4-benzoyl, h-bpa-oh, h-p-bz-phe-oh
(2S)-2-amino-3-(4-benzoylphenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only