CAS RN 104504-45-2
IUPAC Name:
(2S)-2-amino-3-(4-benzoylphenyl)propanoic acid
Synonyms:
para-(benzoyl)-phenylalanine
Molecular Weight (g/mol):
269.3
Molecular Formula:
C16H15NO3
InChi Key:
TVIDEEHSOPHZBR-AWEZNQCLSA-N
SMILES:
N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
4-Benzoyl-L-phenylalanine, 95%
4-Benzoyl-L-phenylalanine, 95%
CAS: 104504-45-2 Molecular Formula: C16H15NO3 Molecular Weight (g/mol): 269.30 MDL Number: MFCD00238102 InChI Key: TVIDEEHSOPHZBR-AWEZNQCLSA-N Synonym: s-2-amino-3-4-benzoylphenyl propanoic acid,l-4-benzoylphenylalanine,para-benzoyl-phenylalanine,h-bpa-oh h-phe 4-bz-oh,4-benzoyl-l-phenylalanine,4-benzoylphenylalanine,l-p-benzoylphenylalanine,phenylalanine, 4-benzoyl,h-bpa-oh,h-p-bz-phe-oh PubChem CID: 7020128 IUPAC Name: (2S)-2-amino-3-(4-benzoylphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O