missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoyl-L-phenylalanine, 95%

Catalog No. AAH52083MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52083MD 250 mg
AAH5208303 1 g
AAH5208306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52083MD Supplier Thermo Scientific Chemicals Supplier No. H52083MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104504-45-2 | C16H15NO3 | 269.30 g/mol

4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 104504-45-2
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.30
MDL Number MFCD00238102
InChI Key TVIDEEHSOPHZBR-AWEZNQCLSA-N
Synonym s-2-amino-3-4-benzoylphenyl propanoic acid, l-4-benzoylphenylalanine, para-benzoyl-phenylalanine, h-bpa-oh h-phe 4-bz-oh, 4-benzoyl-l-phenylalanine, 4-benzoylphenylalanine, l-p-benzoylphenylalanine, phenylalanine, 4-benzoyl, h-bpa-oh, h-p-bz-phe-oh
PubChem CID 7020128
IUPAC Name (2S)-2-amino-3-(4-benzoylphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 269.3
Percent Purity 95%
Chemical Name or Material 4-Benzoyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only