Learn More
4-Azaphthalide, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 5657-51-2 |
|---|---|
| Molecular Formula | C7H5NO2 |
| Molecular Weight (g/mol) | 135.12 |
| InChI Key | YSJHADWSLVFGGT-UHFFFAOYSA-N |
| Synonym | furo 3,4-b pyridin-5 7h-one, 4-azaphthalide, 7h-furo 3,4-b pyridin-5-one, ksc269g5r, 5h,7h-furo 3,4-b pyridin-5-one |
| PubChem CID | 819439 |
| IUPAC Name | 7H-furo[3,4-b]pyridin-5-one |
| SMILES | C1C2=C(C=CC=N2)C(=O)O1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC458450010
|
Thermo Scientific Chemicals
458450010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC458450050
|
Thermo Scientific Chemicals
458450050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC458450250
|
Thermo Scientific Chemicals
458450250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 5657-51-2 | |
| 135.12 | |
| furo 3,4-b pyridin-5 7h-one, 4-azaphthalide, 7h-furo 3,4-b pyridin-5-one, ksc269g5r, 5h,7h-furo 3,4-b pyridin-5-one | |
| 7H-furo[3,4-b]pyridin-5-one |
| C7H5NO2 | |
| YSJHADWSLVFGGT-UHFFFAOYSA-N | |
| 819439 | |
| C1C2=C(C=CC=N2)C(=O)O1 |
Specifications
| 5657-51-2 | |
| 97.5% min. (HPLC) | |
| C7H5NO2 | |
| furo 3,4-b pyridin-5 7h-one, 4-azaphthalide, 7h-furo 3,4-b pyridin-5-one, ksc269g5r, 5h,7h-furo 3,4-b pyridin-5-one | |
| C1C2=C(C=CC=N2)C(=O)O1 | |
| 135.12 | |
| 135.12 | |
| 4-Azaphthalide |
| Authentic | |
| Glass Bottle | |
| 1g | |
| YSJHADWSLVFGGT-UHFFFAOYSA-N | |
| 7H-furo[3,4-b]pyridin-5-one | |
| 819439 | |
| 98% |
Safety and Handling
GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only