CAS RN 5657-51-2
IUPAC Name:
5H,7H-furo[3,4-b]pyridin-5-one
Synonyms:
7H-furo[3,4-b]pyridin-5-one
Molecular Weight (g/mol):
135.12
Molecular Formula:
C7H5NO2
InChi Key:
YSJHADWSLVFGGT-UHFFFAOYSA-N
SMILES:
O=C1OCC2=C1C=CC=N2
1
–
1
of
1
results
1
Promotions Available
4-Azaphthalide, 98%, Thermo Scientific Chemicals
4-Azaphthalide, 98%, Thermo Scientific Chemicals
CAS: 5657-51-2 Molecular Formula: C7H5NO2 Molecular Weight (g/mol): 135.12 InChI Key: YSJHADWSLVFGGT-UHFFFAOYSA-N Synonym: furo 3,4-b pyridin-5 7h-one,4-azaphthalide,7h-furo 3,4-b pyridin-5-one,ksc269g5r,5h,7h-furo 3,4-b pyridin-5-one PubChem CID: 819439 IUPAC Name: 7H-furo[3,4-b]pyridin-5-one SMILES: C1C2=C(C=CC=N2)C(=O)O1