missing translation for 'onlineSavingsMsg'
Learn More

4-Azaindole, 97%

Catalog No. p-7055605
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6422403 1 g
AAH6422406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6422403 Supplier Thermo Scientific Chemicals Supplier No. H6422403

CAS: 272-49-1 | C7H6N2 | 118.14 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol and chloroform. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 272-49-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00971977
InChI Key XWIYUCRMWCHYJR-UHFFFAOYSA-N
Synonym 4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx
PubChem CID 9226
IUPAC Name 1H-pyrrolo[3,2-b]pyridine
SMILES N1C=CC2=NC=CC=C12

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Beilstein 109683
Solubility Information Soluble in methanol and chloroform. Slightly soluble in water.
Formula Weight 118.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Azaindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UY8715000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only