CAS RN 272-49-1
IUPAC Name:
1H-pyrrolo[3,2-b]pyridine
Synonyms:
4-azaindole
Molecular Weight (g/mol):
118.14
Molecular Formula:
C7H6N2
InChi Key:
XWIYUCRMWCHYJR-UHFFFAOYSA-N
SMILES:
N1C=CC2=NC=CC=C12
1
–
1
of
1
results
1
1H-Pyrrolo[3,2-b]pyridine 98.0+%, TCI America™
CAS: 272-49-1 Molecular Formula: C7H6N2 Molecular Weight (g/mol): 118.14 MDL Number: MFCD00971977 InChI Key: XWIYUCRMWCHYJR-UHFFFAOYSA-N Synonym: 4-azaindole,1h-pyrrolo 3,2-b pyridine,1,4-diazaindene,pyrrolopyridine,4h-pyrrolo 3,2-b pyridine,1h-pyrrolo 3.2-b pyridine,zlchem 17,pubchem7675,acmc-1clkx PubChem CID: 9226 IUPAC Name: 1H-pyrrolo[3,2-b]pyridine SMILES: N1C=CC2=NC=CC=C12