missing translation for 'onlineSavingsMsg'
Learn More

4-Amyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7117018
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A070725ML 25 mL
A0707500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A070725ML Supplier TCI America Supplier No. A070725ML

Chemical Identifiers

CAS 5736-91-4
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00014135
InChI Key YAPVGSXODFOBBR-UHFFFAOYSA-N
Synonym 4-pentyloxy benzaldehyde, 4-amyloxybenzaldehyde, p-pentyloxybenzaldehyde, benzaldehyde, 4-pentyloxy, 4-pentyloxybenzaldehyde, benzaldehyde, p-pentyloxy, p-amyloxy benzaldehyde, 4-n-amyloxybenzaldehyde, 4-n-pentyloxybenzaldehyde, p-pentyloxy benzaldehyde
PubChem CID 79815
IUPAC Name 4-pentoxybenzaldehyde
SMILES CCCCCOC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 mL
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Amyloxybenzaldehyde
EINECSNumber (3)-2921
TSCA Yes