missing translation for 'onlineSavingsMsg'
Learn More

4-Amylbenzoic Acid 98.0+%, TCI America™

Catalog No. A0741250G
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0741250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0741250G Supplier TCI America Supplier No. A0741250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26311-45-5
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00002572
InChI Key CWYNKKGQJYAHQG-UHFFFAOYSA-N
Synonym benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
PubChem CID 33479
IUPAC Name 4-pentylbenzoic acid
SMILES CCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 89°C
Color White
Quantity 250 g
Formula Weight 192.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amylbenzoic Acid
EINECSNumber (3)-1338
RTECSNumber DH6273000
TSCA Yes