missing translation for 'onlineSavingsMsg'
Learn More

4-Aminothiophenol, 97%

Catalog No. AAA1408206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1408206 5 g
AAA1408214 25 g
AAA1408222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1408206 Supplier Thermo Scientific Chemicals Supplier No. A1408206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1193-02-8 | C6H6NS | 124.18 g/mol

4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

Solubility
Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.

Chemical Identifiers

CAS 1193-02-8
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007880
InChI Key WCDSVWRUXWCYFN-UHFFFAOYSA-M
Synonym 4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan
PubChem CID 14510
IUPAC Name 4-aminobenzenethiol
SMILES NC1=CC=C([S-])C=C1

Specifications

Melting Point 37°C to 42°C
Boiling Point 156°C to 157°C
Flash Point >110°C (230°F)
Assay Percent Range 97%
Quantity 5 g
UN Number UN3263
Beilstein 906904
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 125.19
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 4-Aminothiophenol
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 214-763-4
RTECSNumber DC0602500
TSCA No
storageNote1 Air Sensitive
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only