missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminostyrene 95.0+%, TCI America™
Supplier: TCI America A101925G
Specifications
| 4-Aminostyrene | |
| White-Yellow | |
| MFCD00015329 | |
| 3531 | |
| LBSXSAXOLABXMF-UHFFFAOYSA-N | |
| 4-ethenylaniline | |
| 73700 | |
| ≥95.0% (T) |
| 1520-21-4 | |
| C8H9N | |
| 25 g | |
| 4-vinylaniline, 4-aminostyrene, p-aminostyrene, benzenamine, 4-ethenyl, 4-aminostryene, ccris 4326, benzenamine,4-ethenyl, p-vinylaniline, p-amino styrene, para-aminostyrene | |
| C=CC1=CC=C(C=C1)N | |
| 119.167 | |
| 119.17 | |
| Crystalline Lumps |
Chemical Identifiers
| 1520-21-4 | |
| 119.167 | |
| LBSXSAXOLABXMF-UHFFFAOYSA-N | |
| 4-ethenylaniline |
| C8H9N | |
| MFCD00015329 | |
| 73700 | |
| C=CC1=CC=C(C=C1)N |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer