missing translation for 'onlineSavingsMsg'
Learn More

4-Aminostyrene 95.0+%, TCI America™

Catalog No. A10195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10195G 5 g
A101925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10195G Supplier TCI America Supplier No. A10195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1520-21-4
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00015329
InChI Key LBSXSAXOLABXMF-UHFFFAOYSA-N
PubChem CID 73700
IUPAC Name 4-ethenylaniline
SMILES C=CC1=CC=C(C=C1)N

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3531
Synonym 4-vinylaniline, 4-aminostyrene, p-aminostyrene, benzenamine, 4-ethenyl, 4-aminostryene, ccris 4326, benzenamine,4-ethenyl, p-vinylaniline, p-amino styrene, para-aminostyrene
Formula Weight 119.17
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Aminostyrene
TSCA Yes
Recommended Storage Freezer