missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminosalicylic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 65-49-6 |
| Linear Formula | H2 NC6H3-2-(OH)CO2 H |
| MDL Number | MFCD00007789 |
| Melting Point | 135°C to 145°C |
| Synonym | 4-Aminosalicylic acid; 4-Amino-2-hydroxybenzoic acid; PAS |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 153.14 |
| Grade | Certified Reference Material |