missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoquinoline, 95+%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 4-Aminoquinoline |
| Point d’ébullition | 332.0°C to 337.0°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 95% min. (HPLC) |
| Quantité | 250mg |
| Renseignements sur la solubilité | Solubility in water: slightly soluble. |
| Poids de formule | 144.17 |
| Pourcentage de pureté | 95+% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
RUO – Research Use Only