accessibility menu, dialog, popup

UserName

NH2N

CAS RN 578-68-7

IUPAC Name: quinolin-4-amine
Synonyms: 4-aminoquinoline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: FQYRLEXKXQRZDH-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)