missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoquinazoline, 97%, Thermo Scientific Chemicals

Catalog No. AAH6694803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6694803 1 g
AAH6694806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6694803 Supplier Thermo Scientific Chemicals Supplier No. H6694803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15018-66-3
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00092016
InChI Key DRYRBWIFRVMRPV-UHFFFAOYSA-N
Synonym quinazolin-4-ylamine, 4-quinazolinamine, 4-aminoquinazoline, 4-quinazolineamine, quinazoline, 4-amino, quinazoline-4-ylamine, 1lq, 4-amino quinazoline, 4-amino-quinazoline, 4-quinazolinamine #
PubChem CID 84759
IUPAC Name quinazolin-4-amine
SMILES NC1=C2C=CC=CC2=NC=N1

Specifications

Quantity 1 g
Beilstein 118621
Formula Weight 145.06
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Aminoquinazoline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber VA1236000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only