accessibility menu, dialog, popup

UserName

CAS RN 15018-66-3

IUPAC Name: quinazolin-4-amine
Synonyms: 4-quinazolinamine
Molecular Weight (g/mol): 145.17
Molecular Formula: C8H7N3
InChi Key: DRYRBWIFRVMRPV-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results