Learn More
4-Aminopyrimidine, 98%
CAS: 591-54-8 | C4H5N3 | 95.105 g/mol
$115.47 - $951.49
Chemical Identifiers
| CAS | 591-54-8 |
|---|---|
| Molecular Formula | C4H5N3 |
| Molecular Weight (g/mol) | 95.105 |
| MDL Number | MFCD00006112 |
| InChI Key | OYRRZWATULMEPF-UHFFFAOYSA-N |
| Synonym | 4-aminopyrimidine, 4-pyrimidinamine, 4-pyrimidineamine, pyrimidin-4-ylamine, pyrimidine, 4-amino, 6-aminopyrimidine, 4-amino pyrimidine, pyrimidine-4-ylamine, pyrimidin-4-yl-amine, 4-pyrimidinamine 9ci |
| PubChem CID | 68958 |
| ChEBI | CHEBI:38616 |
| IUPAC Name | pyrimidin-4-amine |
| SMILES | C1=CN=CN=C1N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH27367MD
|
Thermo Scientific Chemicals
H27367MD |
250 mg |
Each for $115.47
|
|
|||||
|
AAH2736703
|
Thermo Scientific Chemicals
H2736703 |
1 g |
Each for $245.38
|
|
|||||
|
AAH2736706
|
Thermo Scientific Chemicals
H2736706 |
5 g |
Each for $951.49
|
|
|||||
Description
4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 591-54-8 | |
| 95.105 | |
| OYRRZWATULMEPF-UHFFFAOYSA-N | |
| 68958 | |
| pyrimidin-4-amine |
| C4H5N3 | |
| MFCD00006112 | |
| 4-aminopyrimidine, 4-pyrimidinamine, 4-pyrimidineamine, pyrimidin-4-ylamine, pyrimidine, 4-amino, 6-aminopyrimidine, 4-amino pyrimidine, pyrimidine-4-ylamine, pyrimidin-4-yl-amine, 4-pyrimidinamine 9ci | |
| CHEBI:38616 | |
| C1=CN=CN=C1N |
Specifications
| 591-54-8 | |
| C4H5N3 | |
| 250 mg | |
| Slightly soluble in water. | |
| C1=CN=CN=C1N | |
| 95.105 | |
| CHEBI:38616 | |
| 98% |
| 147°C to 152°C | |
| MFCD00006112 | |
| 4-aminopyrimidine, 4-pyrimidinamine, 4-pyrimidineamine, pyrimidin-4-ylamine, pyrimidine, 4-amino, 6-aminopyrimidine, 4-amino pyrimidine, pyrimidine-4-ylamine, pyrimidin-4-yl-amine, 4-pyrimidinamine 9ci | |
| OYRRZWATULMEPF-UHFFFAOYSA-N | |
| pyrimidin-4-amine | |
| 68958 | |
| 95.11 | |
| 4-Aminopyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only