accessibility menu, dialog, popup

UserName

NH2NN

CAS RN 591-54-8

IUPAC Name: pyrimidin-4-amine
Synonyms: pyrimidin-4-amine
Molecular Weight (g/mol): 95.11
Molecular Formula: C4H5N3
InChi Key: OYRRZWATULMEPF-UHFFFAOYSA-N
SMILES: NC1=CC=NC=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)