missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyrimidine, 98%

Catalog No. p-3735497
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC345070010 1 g Glass bottle
AC345070050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC345070010 Supplier Thermo Scientific Chemicals Supplier No. 345070010

CAS: 591-54-8 | C4H5N3 | 95.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-54-8
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.1
MDL Number MFCD00006112
InChI Key OYRRZWATULMEPF-UHFFFAOYSA-N
Synonym 4-aminopyrimidine, 4-pyrimidinamine, 4-pyrimidineamine, pyrimidin-4-ylamine, pyrimidine, 4-amino, 6-aminopyrimidine, 4-amino pyrimidine, pyrimidine-4-ylamine, pyrimidin-4-yl-amine, 4-pyrimidinamine 9ci
PubChem CID 68958
ChEBI CHEBI:38616
IUPAC Name pyrimidin-4-amine
SMILES C1=CN=CN=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 151°C to 155°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 95.1
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4-Aminopyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only