missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopiperidine 96.0+%, TCI America™

Catalog No. p-7117538
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15915ML 5 mL
A159125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15915ML Supplier TCI America Supplier No. A15915ML
Only null left

Chemical Identifiers

CAS 13035-19-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD02179399
InChI Key BCIIMDOZSUCSEN-UHFFFAOYSA-N
Synonym 4-aminopiperidine, 4-piperidinamine, 4-piperidineamine, piperidin-4-ylamine, 4-amino piperidine, 4-piperidylamine, 4-piperidinamine dihydrochloride, pperdn-4-amne, 4-amino, 4-amino-piperidine
PubChem CID 424361
IUPAC Name piperidin-4-amine
SMILES C1CNCCC1N

Specifications

Boiling Point 36°C
Quantity 5 mL
UN Number 2925
Formula Weight 100.17
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Aminopiperidine