missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopiperidine, 95%

Catalog No. AAL2012706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2012706 5 g
AAL2012714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2012706 Supplier Thermo Scientific Chemicals Supplier No. L2012706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13035-19-3 | C5H12N2 | 100.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13035-19-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD02179399
InChI Key BCIIMDOZSUCSEN-UHFFFAOYSA-N
Synonym 4-aminopiperidine, 4-piperidinamine, 4-piperidineamine, piperidin-4-ylamine, 4-amino piperidine, 4-piperidylamine, 4-piperidinamine dihydrochloride, pperdn-4-amne, 4-amino, 4-amino-piperidine
PubChem CID 424361
IUPAC Name piperidin-4-amine
SMILES C1CNCCC1N

Specifications

Density 0.954
Boiling Point 65°C (18 mmHg)
Flash Point 46°C (114°F)
Refractive Index 1.491
Quantity 5 g
UN Number UN2733
Beilstein 1105
Sensitivity Air sensitive; Hygroscopic; May form a precipitate
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 4-Aminopiperidine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only