missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalonitrile 98.0+%, TCI America™

Catalog No. p-7117280
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
A11665G 5 g
A116625G 25 g
2 options

Catalog No. A11665G

Supplier: TCI America A11665G

Only null left

Chemical Identifiers

CAS 56765-79-8
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.149
MDL Number MFCD00041741
InChI Key RRCAJFYQXKPXOJ-UHFFFAOYSA-N
Synonym 4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3
PubChem CID 92529
IUPAC Name 4-aminobenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1N)C#N)C#N

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 143.15
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophthalonitrile