missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalonitrile, 98%

Catalog No. AAL1343303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1343303 1 g
AAL1343306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1343303 Supplier Thermo Scientific Chemicals Supplier No. L1343303

CAS: 56765-79-8 | C8H5N3 | 143.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56765-79-8
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.149
MDL Number MFCD00041741
InChI Key RRCAJFYQXKPXOJ-UHFFFAOYSA-N
Synonym 4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3
PubChem CID 92529
IUPAC Name 4-aminobenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1N)C#N)C#N

Specifications

Melting Point 179°C to 182°C
Quantity 1 g
UN Number UN3439
Beilstein 2937382
Formula Weight 143.15
Percent Purity 98%
Chemical Name or Material 4-Aminophthalonitrile
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 260-370-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only