missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalonitrile, 97%

Catalog No. AC401020100
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401020100 10 g Glass bottle
AC401020250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401020100 Supplier Thermo Scientific Chemicals Supplier No. 401020100
Only null left

CAS: 56765-79-8 | C8H5N3 | 143.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56765-79-8
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.15
MDL Number MFCD00041741
InChI Key RRCAJFYQXKPXOJ-UHFFFAOYSA-N
Synonym 4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3
PubChem CID 92529
IUPAC Name 4-aminobenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1N)C#N)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 177.0°C to 182.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 05,
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 143.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophthalonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a po
EINECSNumber 260-370-6

RUO – Research Use Only