missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalic Acid 95.0+%, TCI America™

Catalog No. p-7117487
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15125G 5 g
A151225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15125G Supplier TCI America Supplier No. A15125G
Only null left

Chemical Identifiers

CAS 5434-21-9
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00013985
InChI Key OXSANYRLJHSQEP-UHFFFAOYSA-N
Synonym 4-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-amino, 4-amino-1,2-benzenedicarboxylic acid, 4-amino phthalic acid, contains 3-aminophthalic acid, pubchem16853, acmc-1arqu, 4-aminophthalic acid, ksc270i4n, ghl.pd_mitscher_leg0.1208
PubChem CID 72912
IUPAC Name 4-aminophthalic acid
SMILES C1=CC(=C(C=C1N)C(=O)O)C(=O)O

Specifications

Melting Point 344°C
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophthalic Acid