missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylarsonic Acid 98.0+%, TCI America™

Catalog No. A053025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A053025G 25 g
A0530500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A053025G Supplier TCI America Supplier No. A053025G

Chemical Identifiers

CAS 98-50-0
Molecular Formula C6H8AsNO3
Molecular Weight (g/mol) 217.056
MDL Number MFCD00007819
InChI Key XKNKHVGWJDPIRJ-UHFFFAOYSA-N
Synonym p-arsanilic acid, arsanilic acid, 4-arsanilic acid, 4-aminophenyl arsonic acid, 4-aminobenzenearsonic acid, atoxylic acid, p-aminobenzenearsonic acid, aminophenylarsine acid, p-anilinearsonic acid, pro-gen
PubChem CID 7389
ChEBI CHEBI:49477
IUPAC Name (4-aminophenyl)arsonic acid
SMILES C1=CC(=CC=C1N)[As](=O)(O)O

Specifications

Quantity 25 g
UN Number 3465
Formula Weight 217.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenylarsonic Acid
RTECSNumber CF7875000
TSCA Yes