missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetonitrile, 99%

Catalog No. p-7081361
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1130406 5 g
AAA1130414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1130406 Supplier Thermo Scientific Chemicals Supplier No. A1130406
Only null left

CAS: 3544-25-0 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3544-25-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00007912
InChI Key YCWRFIYBUQBHJI-UHFFFAOYSA-N
Synonym 4-aminobenzyl cyanide, 4-aminophenylacetonitrile, 2-4-aminophenyl acetonitrile, 4-aminophenyl acetonitrile, p-aminobenzyl cyanide, benzeneacetonitrile, 4-amino, 4-aminobenzylcyanide, 4-amino-phenyl-acetonitrile, 2-4-aminophenyl ethanenitrile, 4-aminobenzyl
PubChem CID 77000
IUPAC Name 2-(4-aminophenyl)acetonitrile
SMILES NC1=CC=C(CC#N)C=C1

Specifications

Melting Point 42°C to 46°C
Boiling Point 312°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3439
Beilstein 1100108
Formula Weight 132.17
Percent Purity 99%
Chemical Name or Material 4-Aminophenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 222-587-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only