missing translation for 'onlineSavingsMsg'
Learn More

(4-Aminophenyl)phosphonic Acid 98.0+%, TCI America™

Catalog No. A3161200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3161200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3161200MG Supplier TCI America Supplier No. A3161200MG
Only null left

Chemical Identifiers

CAS 5337-17-7
Molecular Formula C6H8NO3P
Molecular Weight (g/mol) 173.11
MDL Number MFCD00010580
InChI Key OAOBMEMWHJWPNA-UHFFFAOYSA-N
PubChem CID 53590
IUPAC Name (4-aminophenyl)phosphonic acid
SMILES NC1=CC=C(C=C1)P(O)(O)=O

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 200 mg
Formula Weight 173.11
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Aminophenyl)phosphonic Acid