missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenol hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014417
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014417 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014417 Supplier Merck Emd Millipore Supplier No. PHR1745500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 4-Aminophenol hydrochloride can be used to study cell biology, bioactive small molecules, chemical synthesis, organic building blocks, oxygen compounds and phenols. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 51-78-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula H2 NC6H4 OH · HCl
MDL Number MFCD00012996
Melting Point 300°C to 305°C
Synonym 4-Aminophenol hydrochloride; 4-Hydroxyaniline hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 145.59
Grade Certified Reference Material