missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)pyridine, 97+%

Catalog No. AAL0634514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0634514 25 g
AAL0634522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0634514 Supplier Thermo Scientific Chemicals Supplier No. L0634514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3731-53-1 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3731-53-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006449
InChI Key TXQWFIVRZNOPCK-UHFFFAOYSA-N
Synonym 4-pyridinemethaneamine, 4-aminomethyl pyridine, 4-pyridinemethanamine, 4-pyridylmethylamine, 4-picolylamine, 4-aminomethylpyridine, 4-picolinylamine, 4-pyridinemethylamine, 1-pyridin-4-ylmethanamine, 4-picolinamine
PubChem CID 77317
IUPAC Name pyridin-4-ylmethanamine
SMILES C1=CN=CC=C1CN

Specifications

Melting Point -8°C
Density 1.069
Boiling Point 230°C
Flash Point 108°C (226°F)
Odor Amine-like
Refractive Index 1.5515
Quantity 25 g
UN Number UN2735
Beilstein 108057
Sensitivity Air Sensitive
Formula Weight 108.14
Percent Purity ≥97%
Chemical Name or Material 4-(Aminomethyl)pyridine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 223-092-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only