missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzoic Acid 97.0+%, TCI America™

Catalog No. A096525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A096525G 25 g
A0965100G 100 g
A0965500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A096525G Supplier TCI America Supplier No. A096525G

Chemical Identifiers

CAS 56-91-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00010203
InChI Key QCTBMLYLENLHLA-UHFFFAOYSA-N
Synonym 4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
PubChem CID 65526
IUPAC Name 4-(aminomethyl)benzoic acid
SMILES C1=CC(=CC=C1CN)C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Aminomethyl)benzoic Acid
EINECSNumber (9)-0109
TSCA No