Learn More
4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 95%
CAS: 850568-55-7 | C13H21BClNO2 | 269.576 g/mol
$131.68 - $250.67
Chemical Identifiers
| CAS | 850568-55-7 |
|---|---|
| Molecular Formula | C13H21BClNO2 |
| Molecular Weight (g/mol) | 269.576 |
| MDL Number | MFCD02179455 |
| InChI Key | KPECMJIHZZWTJN-UHFFFAOYSA-N |
| Synonym | 4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl |
| PubChem CID | 16427088 |
| IUPAC Name | [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine;hydrochloride |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52691MD
|
Thermo Scientific Chemicals
H52691MD |
250 mg |
Each for $131.68
|
|
|||||
|
AAH5269103
|
Thermo Scientific Chemicals
H5269103 |
1 g |
Each for $250.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 850568-55-7 | |
| 269.576 | |
| KPECMJIHZZWTJN-UHFFFAOYSA-N | |
| 16427088 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl |
| C13H21BClNO2 | |
| MFCD02179455 | |
| 4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl | |
| [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine;hydrochloride |
Specifications
| 850568-55-7 | |
| C13H21BClNO2 | |
| 250 mg | |
| KPECMJIHZZWTJN-UHFFFAOYSA-N | |
| [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine;hydrochloride | |
| 16427088 | |
| 95% |
| 228°C to 238°C | |
| MFCD02179455 | |
| 4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl | |
| 269.576 | |
| 269.58 | |
| 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only