missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)-2-methoxyphenol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117843
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23305G 5 g
A233025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23305G Supplier TCI America Supplier No. A23305G
Only null left

Chemical Identifiers

CAS 10-2-7149
Molecular Formula C8H11ClNO2
Molecular Weight (g/mol) 188.63
MDL Number MFCD00012864
InChI Key LOYPVODLNGWOLM-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methoxybenzylamine hydrochloride, 4-aminomethyl-2-methoxyphenol hydrochloride, vanillylamine hydrochloride, n-vanillylamine hydrochloride, 2-vanillylamine hydrochloride, 4-hydroxy-3-methoxybenzylamine hcl, phenol, 4-aminomethyl-2-methoxy-, hydrochloride, 4-aminomethyl-2-methoxyphenol, chloride, vanillylamine hcl
PubChem CID 165576
IUPAC Name 4-(aminomethyl)-2-methoxyphenol;hydrochloride
SMILES COC1=C(C=CC(=C1)CN)O.Cl

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
Formula Weight 189.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Aminomethyl)-2-methoxyphenol Hydrochloride