missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyl-1-Boc-4-hydroxypiperidine, 97%

Catalog No. p-7054692
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6307903 1 g
AAH6307906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6307903 Supplier Thermo Scientific Chemicals Supplier No. H6307903
Only null left

CAS: 392331-66-7 | C11H22N2O3 | 230.308 g/mol

4-Aminomethyl-1-Boc-4-hydroxypiperidine can be used in agrochemical, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminomethyl-1-Boc-4-hydroxypiperidine can be used in agrochemical, pharmaceutical and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 392331-66-7
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
MDL Number MFCD11579803
InChI Key XYWCDAFPRBDRER-UHFFFAOYSA-N
PubChem CID 23396319
IUPAC Name tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)(CN)O

Specifications

Color White
Quantity 1 g
Sensitivity Air Sensitive
Synonym tert-butyl 4-aminomethyl-4-hydroxypiperidine-1-carboxylate, 1-boc-4-aminomethyl-4-hydroxypiperidine, tert-butyl 4-aminomethyl-4-hydroxytetrahydro-1 2h-pyridinecarboxylate, 4-aminomethyl-1-boc-4-hydroxypiperidine, 4-aminomethyl-1-boc-piperidin-4-ol, tert-butyl 4-aminomethyl-4-hydroxypiperidinecarboxylate, tert-butyl 4-aminomethyl-4-hydroxy-piperidine-1-carboxylate, 4-aminomethyl-4-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl4-aminomethyl-4-hydroxypiperidine-1-carboxylate
Solubility Information Slightly soluble in water.
Formula Weight 230.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Aminomethyl-1-Boc-4-hydroxypiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only