accessibility menu, dialog, popup

UserName

CAS RN 392331-66-7

IUPAC Name: tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate
Synonyms: tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate
Molecular Weight (g/mol): 230.31
Molecular Formula: C11H22N2O3
InChi Key: XYWCDAFPRBDRER-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(O)(CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results