missing translation for 'onlineSavingsMsg'
Learn More

4-Aminodiphenylmethane 97.0+%, TCI America™

Catalog No. A12605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12605G 5 g
A126025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12605G Supplier TCI America Supplier No. A12605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1135-12-2
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00047861
InChI Key WDTRNCFZFQIWLM-UHFFFAOYSA-N
Synonym 4-aminodiphenylmethane, benzenamine, 4-phenylmethyl, 4-benzyl-phenylamine, 4-benzylbenzenamine, 4-benzylphenylamine, 4-benzylaniline, hcl, 4-benzyl aniline, 4-benzyl-aniline, #, p-aminodiphenylmethan
PubChem CID 136914
IUPAC Name 4-benzylaniline
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)N

Specifications

Melting Point 38°C
Quantity 5 g
Formula Weight 183.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminodiphenylmethane