missing translation for 'onlineSavingsMsg'
Learn More

4-Aminocrotonic Acid Hydrochloride 97.0+%, TCI America™

Catalog No. A301110MG
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A301110MG 10 mg
A3011100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A301110MG Supplier TCI America Supplier No. A301110MG
Only null left

Chemical Identifiers

CAS 2126899-84-9
Molecular Formula C4H7NO2
Synonym trans-4-Amino-2-butenoic Acid Hydrochloride

Specifications

Melting Point 212°C
Quantity 10 mg
Formula Weight 137.56
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminocrotonic Acid Hydrochloride