CAS RN 2126899-84-9
CAS RN 2126899-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Aminocrotonic Acid Hydrochloride 97.0+%, TCI America™
CAS: 2126899-84-9 Molecular Formula: C4H7NO2 Synonym: trans-4-Amino-2-butenoic Acid Hydrochloride