missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric Acid 99.0+%, TCI America™

Supplier:  TCI America A0282100G

 View more versions of this product

Catalog No. A0282100G


Only null left

Description

Description

Specifications

Specifications

195°C
56-12-2
C4H9NO2
4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
C(CC(=O)O)CN
100 g
119
103.12
Crystalline Powder
4-Aminobutyric Acid
White
MFCD00008226
BTCSSZJGUNDROE-UHFFFAOYSA-N
4-aminobutanoic acid
103.121
CHEBI:16865
≥99.0% (T)

Chemical Identifiers

56-12-2
103.121
BTCSSZJGUNDROE-UHFFFAOYSA-N
119
4-aminobutanoic acid
C4H9NO2
MFCD00008226
4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
CHEBI:16865
C(CC(=O)O)CN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0114

RTECSNumber : ES6300000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions